missing translation for 'onlineSavingsMsg'
Learn More

Flavine Adenine Dinucleotide Disodium Salt, TRC

Product Code. 30397902
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30397902 250 mg
30397275 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30397902 Supplier Toronto Research Chemicals Supplier No. TRCF371500250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84366-81-4
Molecular Formula C27 H31 N9 O15 P2 . 2 Na
Molecular Weight (g/mol) 829.51
Synonym FAD sodium salt, 5'-Ester with adenosine riboflavin 5'-(trihydrogen diphosphate) sodium salt (1:2)
IUPAC Name disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxy-tetrahydrofuran-2-yl]methoxy-oxido-phosphoryl] [(2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxo-benzo[g]pteridin-10-yl)-2,3,4-trihydroxy-pentyl] phosphate
SMILES [Na+].[Na+].Cc1cc2N=C3C(=O)NC(=O)N=C3N(C[C@H](O)[C@H](O)[C@H](O)COP(=O)([O-])OP(=O)([O-])OC[C@H]4O[C@H]([C@H](O)[C@@H]4O)n5cnc6c(N)ncnc56)c2cc1C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C27H33N9O15P2.2Na/c1-10-3-12-13(4-11(10)2)35(24-18(32-12)25(42)34-27(43)33-24)5-14(37)19(39)15(38)6-48-52(44,45)51-53(46,47)49-7-16-20(40)21(41)26(50-16)36-9-31-17-22(28)29-8-30-23(17)36;;/h3-4,8-9,14-16,19-21,26,37-41H,5-7H2,1-2H3,(H,44,45)(H,46,47)(H2,28,29,30)(H,34,42,43);;/q;2*+1/p-2/t14-,15+,16+,19-,20+,21+,26+;;/m0../s1
Formula Weight 829.121
Chemical Name or Material Flavine-Adenine Dinucleotide Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.