missing translation for 'onlineSavingsMsg'
Learn More

Flavoxate Hydrochloride, TRC

Product Code. 30407969
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30407969 100 mg
30397276 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30407969 Supplier Toronto Research Chemicals Supplier No. TRCF373500100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3717-88-2
Molecular Formula C24 H25 N O4 . Cl H
Molecular Weight (g/mol) 427.92
Synonym 4H-1-Benzopyran-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl-, 2-(1-piperidinyl)ethyl ester, hydrochloride (1:1), 4H-1-Benzopyran-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl-, 2-(1-piperidinyl)ethyl ester, hydrochloride (9CI), 4H-1-Benzopyran-8-carboxylic acid, 3-methyl-4-oxo-2-phenyl-, 2-piperidinoethyl ester, hydrochloride (6CI,7CI,8CI), 1-Piperidineethanol, 3-methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylate (ester), hydrochloride (8CI), 2-(Piperidin-1-yl)ethyl 3-methyl-4-oxo-2-phenyl-4H-chromene-8-carboxylate hydrochloride, Bladderon, DW 61, Flavoxate hydrochloride, Genurin, NSC 114649, Patricin, Peflate, Piperidinoethyl-3-methylflavone-8-carboxylate hydrochloride, Rec 7/0040, Spasmal, Spasuret, Urispas
IUPAC Name 2-piperidin-1-ylethyl 3-methyl-4-oxo-2-phenylchromene-8-carboxylate;hydrochloride
SMILES Cl.CC1=C(Oc2c(cccc2C1=O)C(=O)OCCN3CCCCC3)c4ccccc4

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C24H25NO4.ClH/c1-17-21(26)19-11-8-12-20(23(19)29-22(17)18-9-4-2-5-10-18)24(27)28-16-15-25-13-6-3-7-14-25;/h2,4-5,8-12H,3,6-7,13-16H2,1H3;1H
Formula Weight 427.155
Chemical Name or Material Flavoxate Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.