missing translation for 'onlineSavingsMsg'
Learn More

Flecainide Acetate, 98%, Thermo Scientific™

Sodium channel blocker

Brand:  Thermo Scientific Alfa Aesar J63527.03

Quantity 1g

 View more versions of this product

Product Code. 15435739

  • ONLINE EXCLUSIVE PRICE
    657.00 €
    625.97 € / 1g
    Save 31.03 €
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in dilute acetic acid, dimethyl sulfoxide, ethanol, methanol and water.

Notes
Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

54143-56-5
474.4
RKXNZRPQSOPPRN-UHFFFAOYSA-N
41022
acetic acid;N-(piperidin-2-ylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide
C19H24F6N2O5
MFCD00214290
flecainide acetate, flecainide monoacetate, flecainide acetate salt, n-2-piperidinylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, n-piperidin-2-ylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, tambocor tn, 2,5-bis-2,2,2-trifluoroethoxy-n-2-piperidinylmethyl benzamide acetate, n-2-piperidylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, benzamide, n-2-piperidinylmethyl-2,5-bis 2,2,2-trifluoroethoxy-, monoacetate
CHEBI:5091
CC(=O)O.C1CCNC(C1)CNC(=O)C2=C(C=CC(=C2)OCC(F)(F)F)OCC(F)(F)F

Specifications

54143-56-5
MFCD00214290
14,4098
Soluble in dilute acetic acid,dimethyl sulfoxide,ethanol,methanol and water.
CC(=O)O.C1CCNC(C1)CNC(=O)C2=C(C=CC(=C2)OCC(F)(F)F)OCC(F)(F)F
474.4
CHEBI:5091
98%
C19H24F6N2O5
1g
flecainide acetate, flecainide monoacetate, flecainide acetate salt, n-2-piperidinylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, n-piperidin-2-ylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, tambocor tn, 2,5-bis-2,2,2-trifluoroethoxy-n-2-piperidinylmethyl benzamide acetate, n-2-piperidylmethyl-2,5-bis 2,2,2-trifluoroethoxy benzamide monoacetate, benzamide, n-2-piperidinylmethyl-2,5-bis 2,2,2-trifluoroethoxy-, monoacetate
RKXNZRPQSOPPRN-UHFFFAOYSA-N
acetic acid;N-(piperidin-2-ylmethyl)-2,5-bis(2,2,2-trifluoroethoxy)benzamide
41022
474.4
Flecainide Acetate
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.