Learn More
Sperian™ Honeywell™ V-Maxx™ Ventilated Safety Glasses
Product Code. 12334749
Change view
Click to view available options
Lens Coating:
Anti-fog
Unit Size:
Each
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Coating Unit Size
12334749 Anti-fog Each
11816980 Anti-fog Each
11807080 Anti-fog Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12334749 Supplier Sperian™ Supplier No. 1006194
  • ONLINE EXCLUSIVE PRICE
    14.51 €
    13.15 € / Pack of 1
    Save 1.36 €
    9% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Fits a wide variety of workers and can be worn over prescription eyewear and with half-mask respirators. Sperian™ Honeywell™ V-Maxx™ Ventilated Safety Glasses have a sporty style for increased wearer acceptance.
  • 180° field of vision for distortion-free peripheral vision
  • Rotating strap attachment system provides easier use with hard hats
  • Compliant with splash resistance tests
  • Indirect ventilation prevents overheating
  • Anti-fog clear polycarbonate lenses
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 9001 / 2000
Lens Coating Anti-fog
Lens Tint Clear
Product Type Safety Goggles
For Use With (Application) Impact Passes at 120 m/s
Headband Material Neoprene
No. per Pack 10/Pk.
Ventilation Details Indirect
Material Polycarbonate