Description
FlexAble Biotin Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate Biotin in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.
Specifications
Specifications
| Quantity | 50 Tests |
| Format | Liquid |