missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ FlexAble CoraLite™ 488 Antibody Labeling Kit for Rabbit IgG

Product Code. 17761509 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
10 x 0.5 μg Reactions
200 x 0.5 μg Reactions
50 x 0.5 μg Reactions
Unit Size:
10 reactions
200 reactions
50 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17761509 50 x 0.5 μg Reactions 50 reactions
17741509 10 x 0.5 μg Reactions 10 reactions
17751509 200 x 0.5 μg Reactions 200 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17761509 Supplier Proteintech™ Supplier No. KFA00150

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Antibody Labeling Kit conjugates CoraLite™ 488 into Rabbit IgG antibodies.

FlexAble CoraLite™ 488 Antibody Labeling Kit for Rabbit IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ 488 in any buffer condition to rabbit IgG primary antibodies from any supplier.

  • One labeling reaction requires only 0.5 μg of antibody, regardless of the antibody concentration
  • Label your rabbit IgG antibody in <10 min without any additional equipment

Specifications

Kit Contents 50 μL CoraLite™ 488 FlexLinker for Rabbit IgG, 100 μL FlexQuencher for Rabbit IgG, 500 μL FlexBuffer, Manual, Protocol Magnet
Quantity 50 x 0.5 μg Reactions
Content And Storage 1 Year at -20°C or 6 Months at +4°C, Avoid Light
Excitation/Emission 500 nm/520 nm
Labeling Target IgG
Species Rabbit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.