Learn More
Proteintech FlexAble CoraLite Plus 647 Antibody Labeling Kit for Human IgG
Product Code. 18080716 Shop All Proteintech Products
Click to view available options
Quantity:
10 Tests
100 Tests
50 Tests
Unit Size:
10 reactions
200 reactions
50 reactions
Product Code. 18080716
Brand: Proteintech KFA10610
FlexAble CoraLite Plus 647 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite Plus 647 in any buffer condition to Human IgG primary antibodies from any supplier.
FlexAble CoraLite™ Plus 647 Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ Plus 647 in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.
Specifications
Quantity 10 Tests
Format Liquid