Learn More
Proteintech FlexAble FITC Plus Antibody Labeling Kit for Human IgG
Product Code. p-200050239 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
10 Tests
100 Tests
50 Tests
Unit Size:
10 reactions
200 reactions
50 reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18085155 10 Tests 10 reactions
18030596 100 Tests 200 reactions
18007295 50 Tests 50 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18085155 Supplier Proteintech Supplier No. KFA10910
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
FlexAble FITC Plus Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate FITC Plus in any buffer condition to Human IgG primary antibodies from any supplier.
FlexAble FITC Plus Antibody Labeling Kit for Human IgG is a novel antibody labeling kit that uses an affinity linker to conjugate FITC Plus in any buffer condition to Human IgG primary antibodies from any supplier. One labeling reaction requires only 0.5 μg of antibody - regardless of the antibody concentration. Label your Human IgG antibody in<10 min without any additional equipment.
Specifications
Quantity 10 Tests
Format Liquid