missing translation for 'onlineSavingsMsg'
Learn More

Interscience™ FlexiPump Pro

Product Code. 16234672
Change view
Click to view available options
Flow Rate:
2.5 L/min.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate unitSize
16234672 2.5 L/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16234672 Supplier Interscience™ Supplier No. 562000
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

FlexiPump Pro is a peristaltic pump with enhanced accuracy for the dispensing of culture media, agar and any type of diluent. Compact and ergonomic, it comes with a user-friendly interface. Traceability is guaranteed by printer and USB connection.

  • Compact and easy to use with 3 dispensing modes: continuous, dose (predetermined volume) and multi-dose (predetermined volume, with an adjustable pause between each dose)
  • Includes 32 customizable programs
  • Easily calibrates and picks up in all types of containers
  • Equipped with a powerful motor allowing the distribution of 50 μL to 99.9 L with a flow rate up to 2.5 L/min.
  • Difference in precision between several distributions reaches 1%
  • Complies with ISO 7218 standards

Specifications

Control Type Analog
Display Type LCD
Enclosure Rating IP31
Flow Rate 2.5 L/min.
Motor Speed 10 to 550 rpm
Reversible No
For Use With Sample Bags, Petri Dish, Test Tubes
For Use With (Application) Dispensing Diluent
Dimensions (L x W x H) 16.1 x 33.2 x 20.8 mm
Voltage 100/240 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.