missing translation for 'onlineSavingsMsg'
Learn More

FLI-06, MedChemExpress

Product Code. 16466966
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466966 100 mg 100mg
16482937 50 mg 50mg
16476878 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16466966 Supplier Medchem Express Supplier No. HY15860

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress FLI-06 is an inhibitor of Notch signaling with an EC50 of 2.3 μM.

FLI-06 is an inhibitor of Notch signaling with an EC50 of 2.3 μM.

Specifications

Chemical Name or Material FLI-06
CAS 313967-18-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C25H30N2O5
Quantity 100 mg
Solubility Information DMSO : ≥ 38 mg/mL (86.66 mM)
SMILES O=C(C1=C(C)NC2=C(C(CC(C)(C)C2)=O)C1C3=CC=C([N+]([O-])=O)C=C3)OC4CCCCC4
Molecular Weight (g/mol) 438.52
Formula Weight 438.52
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.