missing translation for 'onlineSavingsMsg'
Learn More

3M™ Doodlebug™ Floor Edge Cleaning Kit

Product Code. 17169350
Change view
Click to view available options
Style:
Handheld Pad Holder
Swivel Head Pad Holder
Unit Size:
6 sets
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Style Unit Size
17169350 Swivel Head Pad Holder 6 sets
17128560 Handheld Pad Holder Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17169350 Supplier 3M™ Doodlebug™ Supplier No. 7100193869
  • ONLINE EXCLUSIVE PRICE
    320.00 €
    205.78 € / 6 sets
    Save 114.22 €
    35% Off
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Convenient handle options provide versatile cleaning solutions. Unique hook and loop bottom holds 3M™ Doodlebug™ Pads, Sponges and Brushes without the use of adhesives or fasteners.

  • Ideal for maintenance of hard to reach areas
  • Efficient and versatile floor cleaning tool
  • Use the included white pad for polishing operations, and the brown pad for stripping
  • Pads are highly durable and easy to clean
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleaning Kit
Color Brown
For Use With (Application) Cleaning
For Use With (Equipment) Doodlebug™ Pads, Sponges and Brushes
Includes (1) Swivel Head Pad Holder, (1) Polishing Floor Pad
Model 6474 K
Quantity 6 Pk.
Style Swivel Head Pad Holder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.