missing translation for 'onlineSavingsMsg'
Learn More

Florisil (100-200 mesh), J.T. Baker™

Product Code. 15588784 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15588784 500 g Poly 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15588784 Supplier Avantor J.T.Baker Supplier No. M36907
  • ONLINE EXCLUSIVE PRICE
    292.00 €
    250.36 € / 500g
    Save 41.64 €
    14% Off
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color White
Physical Form Powder
CAS 1343-88-0
Quantity 500 g
Molecular Formula MgO3Si
Packaging Poly
Synonym Activated magnesium silicate; Synthetic magnesia-silica gel, Synthetic magnesia-silica gel
InChI Key ZADYMNAVLSWLEQ-UHFFFAOYSA-N
Molecular Weight (g/mol) 100.39
Formula Weight 100.38
Chemical Name or Material Florisil (100-200 mesh)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.