missing translation for 'onlineSavingsMsg'
Learn More

Florisil™, 60-100 mesh, for column chromatography

Product Code. 10174830 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic Drum
Unit Size:
1kg
250g
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10174830 500 g Glass Bottle 500g
10358580 250 g Glass Bottle 250g
10793011 1 kg Glass Bottle 1kg
10512904 5 kg Plastic Drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10174830 Supplier Thermo Scientific Acros Supplier No. 205455000
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Florisil, MgO3Si, CAS Number-1343-88-0, 1kg, White, 01,393; 09,223, 100.39, ZADYMNAVLSWLEQ-UHFFFAOYSA-N, 10, 5509, MFCD00078249, 60-100 mesh, for column chromatography, Glass bottle | CAS: 1343-88-0 | MgO3Si | 100.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for column chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1343-88-0
Molecular Formula MgO3Si
Molecular Weight (g/mol) 100.39
MDL Number MFCD00078249
InChI Key ZADYMNAVLSWLEQ-UHFFFAOYSA-N

Specifications

Color White
Physical Form Fine Powder
Quantity 500 g
Fieser 01,393; 09,223
Merck Index 10, 5509
Packaging Glass Bottle
Particle Size Particle size analysis (typical values): 60 mesh: 2.0%, 70 mesh: 20.0%, 80 mesh: 41.0%, 100 mesh: 30.0%, 140 mesh: 6.9%
Formula Weight 100.39
Chemical Name or Material Florisil™, 60-100 mesh, for column chromatography

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.