missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for column chromatography
Specifications
Specifications
| Color | White |
| Physical Form | Fine Powder |
| Quantity | 5 kg |
| Fieser | 01,393; 09,223 |
| Merck Index | 10, 5509 |
| Packaging | Plastic Drum |
| Particle Size | Particle size analysis (typical values): 60 mesh: 2.0%, 70 mesh: 20.0%, 80 mesh: 41.0%, 100 mesh: 30.0%, 140 mesh: 6.9% |
| Formula Weight | 100.39 |
| Chemical Name or Material | Florisil™, 60-100 mesh, for column chromatography |
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?