missing translation for 'onlineSavingsMsg'
Learn More

Florisil™, 60 to 100 Mesh, J.T. Baker™

Product Code. 15578784 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15578784 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15578784 Supplier Avantor J.T.Baker Supplier No. M36807
 Restricted Item
Estimated Shipment: 14-06-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective Adsorbent for Gas Chromatography

Chemical Identifiers

CAS 1343-88-0
Molecular Formula MgO3Si
Molecular Weight (g/mol) 100.39
InChI Key ZADYMNAVLSWLEQ-UHFFFAOYSA-N

Specifications

Quantity 500 g
Mesh Size 60 to 100 Mesh
Packaging Poly Bottle
Specific Gravity 1L = 2.51kg
Chemical Name or Material Florisil™
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.