missing translation for 'onlineSavingsMsg'
Learn More

Florisil™, Activated at 675°C, 60 to 100 Mesh, BAKER ANALYZED™ Reagent, J.T. Baker™

Product Code. 15518124 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Poly Pail
Wide Mouth Amber Glass Bottle
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15518124 2 kg Poly Pail 2kg
15508124 500 g Wide Mouth Amber Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15518124 Supplier Avantor J.T.Baker Supplier No. 337208
  • ONLINE EXCLUSIVE PRICE
    1150.00 €
    990.67 € / 2kg
    Save 159.33 €
    13% Off
 Restricted Item
Estimated Shipment: 16-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

    Suitable for Use in Chromatographic Cleanup of Pesticide Residues

Chemical Identifiers

CAS 1343-88-0
Molecular Formula MgO3Si
Molecular Weight (g/mol) 100.39
InChI Key ZADYMNAVLSWLEQ-UHFFFAOYSA-N

Specifications

Quantity 2 kg
Mesh Size 60 to 100 Mesh
Packaging Poly Pail
Specific Gravity 1L = 2.51kg
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Florisil™, Activated at 675°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.