missing translation for 'onlineSavingsMsg'
Learn More

Florisil™, Honeywell Fluka™

Product Code. 15652080 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15652080 250 g 250g
15642080 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15652080 Supplier Honeywell-Fluka Supplier No. 220736250G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

100-200 mesh

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1343-88-0
Molecular Formula MgO3Si
Molecular Weight (g/mol) 100.39
MDL Number MFCD00078249
InChI Key ZADYMNAVLSWLEQ-UHFFFAOYSA-N

Specifications

Quantity 250 g
Mesh Size 100 to 200mesh
UN Number NONH for all modes of transport
Formula Weight 100.39g/mol
Chemical Name or Material Florisil™
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.