missing translation for 'onlineSavingsMsg'
Learn More

Floxuridine, MedChemExpress

Product Code. 16472006
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472006 500 mg 500mg
16448086 100 mg 100mg
16411376 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16472006 Supplier Medchem Express Supplier No. HYB0097
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Floxuridine (5-Fluorouracil 2'-deoxyriboside) is a pyrimidine analog and known as an oncology antimetabolite. Floxuridine inhibits Poly(ADP-Ribose) polymerase and induces DNA damage by activating the ATM and ATR checkpoint signaling pathways in vitro. Floxuridine is a extreamly potent inhibitor for S. aureus infection and induces cell apoptosis. Floxuridine has antiviral effects against HSV and CMV.

Floxuridine (5-Fluorouracil 2'-deoxyriboside) is a pyrimidine analog and known as an oncology antimetabolite. Floxuridine inhibits Poly(ADP-Ribose) polymerase and induces DNA damage by activating the ATM and ATR checkpoint signaling pathways in vitro. Floxuridine is a extreamly potent inhibitor for S. aureus infection and induces cell apoptosis. Floxuridine has antiviral effects against HSV and CMV.

Specifications

Chemical Name or Material Floxuridine
CAS 50-91-9
Color White
Purity Grade Notes Research
Molecular Formula C9H11FN2O5
Quantity 500 mg
Synonym 5-Fluorouracil 2'-deoxyriboside
Solubility Information DMSO : 250 mg/mL (1015.48 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (203.10 mM)
SMILES OC[C@@H]1[C@H](C[C@H](N2C(NC(C(F)=C2)=O)=O)O1)O
Molecular Weight (g/mol) 246.19
Formula Weight 246.19
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.