missing translation for 'onlineSavingsMsg'
Learn More

Fluazinam, MedChemExpress

Product Code. 16482286
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482286 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16482286 Supplier Medchem Express Supplier No. HYB1839
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fluazinam is a broad spectrum pyridinamine fungal inhibitor.

Fluazinam is a broad spectrum pyridinamine fungal inhibitor.

Specifications

Chemical Name or Material Fluazinam
CAS 79622-59-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C13H4Cl2F6N4O4
Quantity 500 mg
Solubility Information DMSO : 125 mg/mL (268.77 mM; Need ultrasonic)
SMILES FC(C1=CN=C(NC2=C([N+]([O-])=O)C=C(C(F)(F)F)C(Cl)=C2[N+]([O-])=O)C(Cl)=C1)(F)F
Molecular Weight (g/mol) 465.09
Formula Weight 465.09
Percent Purity 98.31%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.