missing translation for 'onlineSavingsMsg'
Learn More

Flubendazole alcohol, TRC

Product Code. 30136721
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136721 1 g 1g
30011659 10 mg 10mg
30010459 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136721 Supplier Toronto Research Chemicals Supplier No. TRCH9424551G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82050-12-2
Molecular Formula C16 H14 F N3 O3
Molecular Weight (g/mol) 315.3
Synonym Carbamic acid, [5-[(4-fluorophenyl)hydroxymethyl]-1H-benzimidazol-2-yl]-, methyl ester (9CI), Reduced-flubendazole, UMF 060
IUPAC Name methyl N-[6-[(4-fluorophenyl)-hydroxymethyl]-1H-benzimidazol-2-yl]carbamate
SMILES COC(=O)Nc1nc2ccc(cc2[nH]1)C(O)c3ccc(F)cc3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H14FN3O3/c1-23-16(22)20-15-18-12-7-4-10(8-13(12)19-15)14(21)9-2-5-11(17)6-3-9/h2-8,14,21H,1H3,(H2,18,19,20,22)
Formula Weight 315.1019
Chemical Name or Material Flubendazole-hydroxy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.