missing translation for 'onlineSavingsMsg'
Learn More

Flucloxacillin Sodium, TRC

Product Code. 30007780
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007780 100 mg 100mg
30006548 1 g 1g
17954375 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007780 Supplier Toronto Research Chemicals Supplier No. TRCF419200100MG
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1847-24-1
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[[3-(2-chloro-6-fluorophenyl)-5-methyl-4-isoxazolyl]carbonyl]amino]-3,3-dimethyl-7-oxo-, sodium salt (1:1), (2S,5R,6R)-∣4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[3-(2-chloro-6-fluorophenyl)-5-methyl-4-isoxazolecarboxamido]-3,3-dimethyl-7-oxo-, sodium salt (7CI)∣4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[[3-(2-chloro-6-fluorophenyl)-5-methyl-4-isoxazolyl]carbonyl]amino]-3,3-dimethyl-7-oxo-, monosodium salt, (2S,5R,6R)- (9CI)∣4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[[[3-(2-chloro-6-fluorophenyl)-5-methyl-4-isoxazolyl]carbonyl]amino]-3,3-dimethyl-7-oxo-, monosodium salt, [2S-(2α,5α,6β)]-∣Floxacillin sodium∣Floxapen∣Floxapen sodium∣Fluclox∣Flucloxacillin sodium∣Flucloxacillin sodium salt∣Monosodium flucloxacillin∣NSC 277175∣Sodium [3-(2-chloro-6-fluorophenyl)-5-methyl-4-isoxazolyl]penicillin∣Sodium flucloxacillin∣Staphylex∣(2S,5R,6R)-6-[[[3-(2-Chloro-6-fluorophenyl)-5-methyl-4-isoxazolyl]carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid sodium salt

Specifications

Format Neat
Molecular Weight (g/mol) 475.85
Product Type Metabolite∣API
Quantity 100 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.