missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flufenoxuron-d3 (2,6-difluorobenzoyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Flufenoxuron D3 (2,6-Difluorobenzoyl D3) |
| Molecular Formula | C21 D3 H8 Cl F6 N2 O3 |
| Quantity | 0.01 g |
| Synonym | Flufenoxuron D3 (2,6-Difluorobenzoyl D3), N-[[[4-[2-Chloro-4-(trifluoromethyl)phenoxy]-2-fluorophenyl]amino]carbonyl]-2,6-difluorobenzamide-3,4,5-d3, Flufenoxuron-d3 |
| InChI Formula | InChI=1 S/C21H11ClF6N2O3/c22-12-8-10(21(26,27)28)4-7-17(12)33-11-5-6-16(15(25)9-11)29-20(32)30-19(31)18-13(23)2-1-3-14(18)24/h1-9 H,(H2,29,30,31,32)/i1D,2 D,3 D |
| SMILES | [2 H]c1c([2 H])c(F)c(C(=O)NC(=O)Nc2ccc(Oc3ccc(cc3Cl)C(F)(F)F)cc2F)c(F)c1[2 H] |
| IUPAC Name | N-[[4-[2-chloro-4-(trifluoromethyl)phenoxy]-2-fluorophenyl]carbamoyl]-3,4,5-trideuterio-2,6-difluorobenzamide |
| Molecular Weight (g/mol) | 491.7855 |
| Formula Weight | 491.0551 g/mol |
| Percent Purity | 98 atom %, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction