Learn More
Thermo Scientific™ Fluid Transfer Assembly
Specialty Fluid Transfer Assembly products integrate easily within your existing process and equipment to maximize productivity, reduce cross-contamination and keep your process running smoothly.
Brand:  Thermo Scientific™ SS00062-I
 View more versions of this product
Product Code. 15969916
  • POA / 25 units
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
Specialized connectors and tubing in our ready-designed assemblies provide an off-the-shelf solution that addresses many fluid transfer needs while delivering efficiency and increasing process integrity.
  • End treatments facilitate simple interconnection with existing process and equipment
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
TRUSTED_SUSTAINABILITY
Specifications
Specifications
91.4cm of 12.7cm ID Silicone Tubing with 3.8cm Sanitary and 1.3cm MPX Body Connections