Learn More
Thermo Scientific™ Fluid Transfer Assembly
Specialty Fluid Transfer Assembly products integrate easily within your existing process and equipment to maximize productivity, reduce cross-contamination and keep your process running smoothly.
Brand: Thermo Scientific™ SS00064-I
Description
Specialized connectors and tubing in our ready-designed assemblies provide an off-the-shelf solution that addresses many fluid transfer needs while delivering efficiency and increasing process integrity.- End treatments facilitate simple interconnection with existing process and equipment
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
Specifications
| 91.4cm of 12.7cm ID Silicone Tubing with 3.8cm Sanitary and 1.3cm MPX insert Connections |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.