Learn More
Thermo Scientific™ Fluid Transfer Assembly, Y Set
Product Code. 17800637 Shop All Thermo Scientific Products
Change view
Click to view available options
Tubing Style:
3 x TPE 374
Connector Types:
Luer
MPX body-insert
MPX insert-body
Dimensions:
1/2 in. ID x 3/4 in. wall
1/4 in. ID x 7/16 in. wall
1/8 in. ID x 1/4 in. wall
3/8 in. ID x 5/8 in. wall
Unit Size:
Pack of 10
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Style Connector Types Dimensions unitSize
17800637 3 x TPE 374 MPX body-insert 1/4 in. ID x 7/16 in. wall Pack of 10
17830687 3 x TPE 374 MPX insert-body 1/4 in. ID x 7/16 in. wall Pack of 10
17810637 3 x TPE 374 MPX insert-body 3/8 in. ID x 5/8 in. wall Pack of 10
17870627 3 x TPE 374 MPX body-insert 1/2 in. ID x 3/4 in. wall Pack of 10
17820637 3 x TPE 374 Luer 1/8 in. ID x 1/4 in. wall Pack of 10
17840687 3 x TPE 374 MPX insert-body 1/2 in. ID x 3/4 in. wall Pack of 10
17890627 3 x TPE 374 MPX body-insert 3/8 in. ID x 5/8 in. wall Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 17800637 Supplier Thermo Scientific™ Supplier No. SH31108.01
  • ONLINE EXCLUSIVE PRICE
    356.00 €
    303.84 € / Pack of 10
    Save 52.16 €
    14% Off
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.
  • One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
TRUSTED_SUSTAINABILITY
Specifications
Dimensions 1/4 in. ID x 7/16 in. wall
Tubing Style 3 x TPE 374
Connector Types MPX body-insert
Product Type Y Set
Tubing Size 2 x 20 in. to 20 in. (Length)