missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Flumazenil

Product Code. 30000705
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30000705 10 mg 10mg
18064952 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30000705 Supplier R&D Systems™ Supplier No. 1328/10
  • ONLINE EXCLUSIVE PRICE
    157.55 €
    141.75 € / 10mg
    Save 15.80 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Benzodiazepine antagonist

Benzodiazepine antagonist, non-selective for α1, α2, α3 or α5-containing GABAA receptors. Centrally active upon systemic administration in vivo. Also inhibits repressor element-1 silencing transcription factor (REST) by targeting degradation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78755-81-4
Molecular Formula C15H14FN3O3

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 303.29
Purity >99%
Molecular Weight (g/mol) 303.293
Chemical Name or Material Flumazenil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.