missing translation for 'onlineSavingsMsg'
Learn More

Flumioxazin, TRC

Product Code. 30133837
Change view
Click to view available options
Quantity:
10 mg
50 mg
1 g
Unit Size:
10mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133837 1 g 1g
17983215 10 mg 10mg
17951285 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133837 Supplier Toronto Research Chemicals Supplier No. TRCF5004701G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103361-09-7
Molecular Formula C19 H15 F N2 O4
Molecular Weight (g/mol) 354.33
Synonym 1H-Isoindole-1,3(2H)-dione, 2-[7-fluoro-3,4-dihydro-3-oxo-4-(2-propynyl)-2H-1,4-benzoxazin-6-yl]-4,5,6,7-tetrahydro- (9CI), 2-[7-Fluoro-3,4-dihydro-3-oxo-4-(2-propyn-1-yl)-2H-1,4-benzoxazin-6-yl]-4,5,6,7-tetrahydro-1H-isoindole-1,3(2H)-dione, 2H-1,4-Benzoxazine, 1H-isoindole-1,3(2H)-dione deriv., 2-(7-Fluoro-3-oxo-4-(prop-2-ynyl)-3,4-dihydro-2H-benzo[b][1,4]oxazin-6-yl)-4,5,6,7-tetrahydro-2H-isoindole-1,3-dione, BroadStar, Chateau, Encompass,, Flumioxazin, Flumizin, Guillotine, Pledge, S 53482, Sumimax, Sumisoya, SureGuard, V 53482, Valor, Valor SX
IUPAC Name 2-(7-fluoro-3-oxo-4-prop-2-ynyl-1,4-benzoxazin-6-yl)-4,5,6,7-tetrahydroisoindole-1,3-dione
SMILES Fc1cc2OCC(=O)N(CC#C)c2cc1N3C(=O)C4=C(CCCC4)C3=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C19H15FN2O4/c1-2-7-21-15-9-14(13(20)8-16(15)26-10-17(21)23)22-18(24)11-5-3-4-6-12(11)19(22)25/h1,8-9H,3-7,10H2
Formula Weight 354.1016
Percent Purity >95%
Chemical Name or Material Flumioxazin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.