missing translation for 'onlineSavingsMsg'
Learn More

Flunixin D3 (methyl D3) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30321946
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30321946 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321946 Supplier Dr. Ehrenstorfer Supplier No. DREA13727010AL100
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Flunixin D3 (methyl D3)
CAS 1015856-60-6
Solvent or Matrix Acetonitrile
Molecular Formula C14 2H3 H8 F3 N2 O2
Quantity 1 mL
Synonym Flunixin D3 (methyl D3)
InChI Formula InChI=1S/C14H11F3N2O2/c1-8-10(14(15,16)17)5-2-6-11(8)19-12-9(13(20)21)4-3-7-18-12/h2-7H,1H3,(H,18,19)(H,20,21)/i1D3
SMILES [2H]C([2H])([2H])c1c(Nc2ncccc2C(=O)O)cccc1C(F)(F)F
IUPAC Name 2-[2-(trideuteriomethyl)-3-(trifluoromethyl)anilino]pyridine-3-carboxylic acid
Molecular Weight (g/mol) 299.26
Formula Weight 299.0961 g/mol
Show More Show Less
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.