missing translation for 'onlineSavingsMsg'
Learn More

Fluocinolone Acetonide Acetate, TRC

Product Code. 30007996
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007996 250 mg 250mg
30008874 100 mg 100mg
30009927 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007996 Supplier Toronto Research Chemicals Supplier No. TRCF455805250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 356-12-7
Molecular Formula C26 H32 F2 O7
Molecular Weight (g/mol) 494.52
Synonym (6alpha,11beta,16alpha)-21-(Acetyloxy)-6,9-difluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione, 6alpha,9-Difluoro-11beta,16alpha,17,21-tetrahydroxypregna-1,4-diene-3,20-dione 21-acetate cyclic 16,17-acetal with acetone, 2H-Naphth[2',1':4,5]indeno[1,2-d][1,3]dioxole pregna-1,4-diene-3,20-dione deriv., Biscosal, Dermaplus, Flucetonide, Flucinar, Fluocinolide, Fluocinolide acetate, Fluocinolone acetonide 21-acetate, Fluocinolone acetonide acetate, Fluocinonide, Lidex, Lidex E, Metosyn, NSC 101791, Straderm, Topsym, Topsymin, Topsyn, Topsyne, Vanos, 6α,9-Difluoro-11β-hydroxy-16α,17-(1-methylethylidenedioxy)-3,20-dioxopregna-1,4-dien-21-yl Acetate
IUPAC Name [2-[(1S,2S,4R,8S,9S,11S,12R,13S,19S)-12,19-difluoro-11-hydroxy-6,6,9,13-tetramethyl-16-oxo-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-dien-8-yl]-2-oxoethyl] acetate
SMILES CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]23C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C26H32F2O7/c1-13(29)33-12-20(32)26-21(34-22(2,3)35-26)10-15-16-9-18(27)17-8-14(30)6-7-23(17,4)25(16,28)19(31)11-24(15,26)5/h6-8,15-16,18-19,21,31H,9-12H2,1-5H3/t15-,16-,18-,19-,21+,23-,24-,25-,26+/m0/s1
Formula Weight 494.2116
Percent Purity >95%
Chemical Name or Material Fluocinolone acetonide acetate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.