missing translation for 'onlineSavingsMsg'
Learn More

Fluoranthene, 98%

Product Code. 10102390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10102390 100 g Glass bottle 100g
10182380 25 g Glass bottle 25g
10646112 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10102390 Supplier Thermo Scientific Acros Supplier No. 119171000
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 206-44-0 | C16H10 | 202.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206-44-0
Molecular Formula C16H10
Molecular Weight (g/mol) 202.26
MDL Number MFCD00001184
InChI Key GVEPBJHOBDJJJI-UHFFFAOYSA-N
Synonym idryl, 1,2-benzacenaphthene, benzo jk fluorene, rcra waste number u120, 1,2-1,8-naphthylene benzene, benzene, 1,2-1,8-naphthalenediyl, 1,2-1,8-naphthalenediyl benzene, benzene, 1,2-1,8-naphthylene, benzo j,k fluorene
PubChem CID 9154
ChEBI CHEBI:33083
IUPAC Name fluoranthene
SMILES C1=CC=C2C(=C1)C1=CC=CC3=CC=CC2=C13

Specifications

Melting Point 109.0°C to 111.0°C
Color Beige-Gray to Green-Yellow or Yellow
Boiling Point 384.0°C
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Beilstein 05, I, 344
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether, acetone and benzene
Formula Weight 202.26
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Fluoranthene, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Fluoranthene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.