missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluoranthene, 98%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 109.0°C to 111.0°C |
| Color | Beige-Gray to Green-Yellow or Yellow |
| Boiling Point | 384.0°C |
| Flash Point | >100°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Quantity | 500 g |
| Beilstein | 05, I, 344 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in ether, acetone and benzene |
| Show More |
Safety and Handling
Product Identifier
- Fluoranthene
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P264-Wash thoroughly after handling.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : fluoranthene
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction