missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluoranthene-d10, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Fluoranthene D10 |
| CAS | 93951-69-0 |
| Molecular Formula | C16 2H10 |
| Quantity | 0.1 g |
| Synonym | Fluoranthene-1,2,3,4,5,6,7,8,9,10-d10, Fluoranthene-d10 (9 CI), Perdeuterated fluoranthene, [10-2 H]Fluoranthene, Fluoranthene-D10 |
| InChI Formula | InChI=1 S/C16H10/c1-2-8-13-12(7-1)14-9-3-5-11-6-4-10-15(13)16(11)14/h1-10 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2c(c1[2 H])c3c([2 H])c([2 H])c([2 H])c4c([2 H])c([2 H])c([2 H])c2c34 |
| IUPAC Name | 1,2,3,4,5,6,7,8,9,10-decadeuteriofluoranthene |
| Molecular Weight (g/mol) | 212.31 |
| Formula Weight | 212.141 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction