missing translation for 'onlineSavingsMsg'
Learn More

Fluoranthene, TRC

Product Code. 30134640
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134640 25 g 25g
30012167 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134640 Supplier Toronto Research Chemicals Supplier No. TRCF46199025G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206-44-0
Molecular Formula C16 H10
Molecular Weight (g/mol) 202.25
Synonym Fluoranthene, 1,2-(1,8-Naphthylene)benzene, Benzene, 1,2-(1,8-naphthalenediyl)-, Benzo[jk]fluorene, ClusterCarbon, Idryl, NSC 6803
IUPAC Name fluoranthene
SMILES c1ccc2c(c1)c3cccc4cccc2c34

Specifications

Quantity 25 g
InChI Formula InChI=1S/C16H10/c1-2-8-13-12(7-1)14-9-3-5-11-6-4-10-15(13)16(11)14/h1-10H
Formula Weight 202.0783
Chemical Name or Material Fluoranthene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.