missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FluoReporter™ lacZ Flow Cytometry Kit

Product Code. 10378542
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10378542 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10378542 Supplier Invitrogen™ Supplier No. F1930
  • ONLINE EXCLUSIVE PRICE
    902.00 €
    640.42 € / Pack of 1
    Save 261.58 €
    29% Off
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

FluoReporter™ lacZ Flow Cytometry Kit

The FluoReporter™ lacZ Flow Cytometry Kit provides materials and protocols for quantitating β-galactosidase activity with fluorescein di-V-galactoside (FDG) in single cells using flow cytometry. This kit is accompanied by a license to practice patented techniques for loading FDG by hypotonic shock and improving retention of fluorescein in lacZ-positive cells.

TRUSTED_SUSTAINABILITY

Specifications

Product Type β-Galactosidase Reporter Gene Assay System
Detection Method Fluorescence
Content And Storage Contains 1 vial FDG (0.5 mL, 20 mM in water/DMSO/ethanol, 8:1:1, v/v), 1 vial PETG (2.0 mL, 50 mM in water), chloroquine diphosphates (1.0 mL, 30 mM in water) and 1 vial of propidium iodide (1.0 mL, 150 μM in water).

Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature
Product Line FluoReporter
Quantity 1 Kit
Form Solution

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.