missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Fluorescamine

Product Code. 16484667
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484667 50 mg 50mg
16428758 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16484667 Supplier MedChemExpress Supplier No. HYD0715
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fluorescamine( Ro 20-7234) is a spiro compound that is not fluorescent itself, but reacts with primary amines to form highly fluorescent products.

Specifications

Quantity 50 mg
Product Type Dye Reagents
Molecular Weight (g/mol) 278.26 g/mol
Solubility DMSO : ≥ 33 mg/mL (118.59 mM)
Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Shipping Condition Room temperature
Target HBV
CAS 38183-12-9
Synonym Ro 20-7234
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.