missing translation for 'onlineSavingsMsg'
Learn More

Fluorescamine, MP Biomedicals™

Product Code. 11471082 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
25mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471082 25 mg 25mg
11481082 100 mg 100mg
11491082 250 mg 250mg
11401092 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11471082 Supplier MP Biomedicals Supplier No. 0219518325
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 38183-12-9
Molecular Formula C17H10O4
Molecular Weight (g/mol) 278.26
MDL Number MFCD00005928
InChI Key ZFKJVJIDPQDDFY-UHFFFAOYNA-N
Synonym 4-phenylspiro[furan-2(3H)-1'-(3'-H)-isobenzofuran]-3, 3'-dione
PubChem CID 37927
SMILES O=C1OC2(OC=C(C2=O)C2=CC=CC=C2)C2=CC=CC=C12

Specifications

IUPAC Name 4'-phenyl-3H,3'H-spiro[2-benzofuran-1,2'-furan]-3,3'-dione
Formula Weight 278.3
Color White
Quantity 25 mg
Physical Form Powder
Chemical Name or Material Fluorescamine, For TLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.