missing translation for 'onlineSavingsMsg'
Learn More

Fluorescamine, pure

Product Code. 10144450 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
2.5 g
Packaging:
Glass bottle
Unit Size:
100mg
2.5g
25mg
500mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10144450 25 mg Glass bottle 25mg
10153730 100 mg Glass bottle 100mg
10551831 500 mg Glass bottle 500mg
11946201 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10144450 Supplier Thermo Scientific Acros Supplier No. 191670250
  • ONLINE EXCLUSIVE PRICE
    56.65 €
    55.00 € / 25mg
    Save 1.65 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 38183-12-9 | C17H10O4 | 278.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38183-12-9
Molecular Formula C17H10O4
Molecular Weight (g/mol) 278.26
MDL Number MFCD00005928
InChI Key ZFKJVJIDPQDDFY-UHFFFAOYNA-N
PubChem CID 37927
IUPAC Name 4'-phenylspiro[2-benzofuran-3,2'-furan]-1,3'-dione
SMILES O=C1OC2(OC=C(C2=O)C2=CC=CC=C2)C2=CC=CC=C12

Specifications

Melting Point 155.0°C to 157.0°C
Solubility Information Solubility in water: .. Other solubilities: soluble in diethyl ether
Infrared Spectrum Authentic
Formula Weight 278.26
Grade Pure
Packaging Glass bottle
Specific Extinction 900 (235nm, ether) min.
Color Yellow
Quantity 25 mg
Physical Form Powder
Chemical Name or Material Fluorescamine
Show More Show Less
compliance-icons
Product Identifier
  • Fluorescamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.