missing translation for 'onlineSavingsMsg'
Learn More

Fluorescamine

Product Code. 11360368
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11360368 500 mg 500mg
11370368 25 mg 25mg
11380368 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11360368 Supplier Thermo Scientific Alfa Aesar Supplier No. 043749.ME
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

For fluorescent detection of amino acids, peptides and proteins | CAS: 38183-12-9 | C17H10O4 | 278.26 g/mol

For fluorescent detection of amino acids, peptides and proteins.Fluorescamine plays an important role as a reagent for the detection of amines, peptides and proteins. It is also used as a substrate for measuring protease activity. It is utilized in the detection and quantitation of residual aminopenicillins and sulfonamides in honey by HPLC. It finds application in the determination of lisinopril in human plasma and urine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38183-12-9
Molecular Formula C17H10O4
Molecular Weight (g/mol) 278.26
MDL Number MFCD00005928
InChI Key ZFKJVJIDPQDDFY-UHFFFAOYNA-N
Synonym fluorescamine, fluram, spiro furan-2 3h ,1' 3'h-isobenzofuran-3,3'-dione, 4-phenyl, 4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dione, 4-phenylspiro furan-2 3h ,1'-phthalan-3,3'-dione, 4-phenylspiro furan-2 3h-1'-3'-h-isobenzofuran-3,3'-dione, zfkjvjidpqddfy-uhfffaoysa, 4'-phenylspiro 2-benzofuran-3,2'-furan-1,3'-dion
PubChem CID 37927
SMILES O=C1OC2(OC=C(C2=O)C2=CC=CC=C2)C2=CC=CC=C12

Specifications

Melting Point 155°C to 158°C
Color White to Cream
Quantity 500 mg
Merck Index 14,4158
Solubility Information Soluble in acetone,ethanol,chloroform,dimethylsulfoxide and acetonitrile. Slightly soluble in water.
IUPAC Name 4'-phenyl-3H,3'H-spiro[2-benzofuran-1,2'-furan]-3,3'-dione
Formula Weight 278.27
Physical Form Powder
Chemical Name or Material Fluorescamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.