missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fluorescein-5-Maleimide

Product Code. 11564786
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 11564786

Brand: Invitrogen™ F150

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Alternative to fluorescein

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Product Type Maleimide
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Chemical Reactivity Thiol
Shipping Condition Room Temperature
Reactive Moiety Maleimide
Label or Dye FITC (Fluorescein)
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.