missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Fluorescein Cadaverine; 5-((5-Aminopentyl)thioureidyl)Fluorescein, Dihydrobromide Salt

Product Code. 11519046
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11519046 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11519046 Supplier Molecular Probes™ Supplier No. A10466

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

5-((5-aminopentyl)thioureidyl)fluorescein, dihydrobromide salt (fluorescein cadaverine)

The primary aliphatic amine of fluorescein cadaverine can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a generate stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3). Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247).
TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mg
Product Type Fluorescein Cadaverine
Content And Storage Store at room temperature and protect from light.
Chemical Reactivity Carboxylic Acid, Ketone, Aldehyde
Shipping Condition Room Temperature
Reactive Moiety Amine, Cadaverine
Label or Dye FITC (Fluorescein)
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.