missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorescein-CREBtide Substrate
Description
A fluorescein-labeled peptide substrate that is based upon the CREB ( cAMP responsive element binding) protein. The peptide substrate is phosphorylated by a wide range of Ser/Thr kinases.
For use in combination with the LanthaScreen™ Tb-CREB [pSer133] antibody (PV3542) to evaluate inhibitors in medium to high throughput screening applications.
For use in combination with the LanthaScreen™ Tb-CREB [pSer133] antibody (PV3542) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
Specifications
| Amino Acid Sequence | Fluorescein- KRREILSRRPSYR |
| Assay Entry | Biochemical terbium phospho-specific activity |
| Content And Storage | 1 mg/mL Fluorescein-CREBtide substrate in water. Store at -20°C and protect from light. |
| Detection Method | Fluorescence |
| Quantity | 1 mg |
| Substrate | Fluorescein-CREBtide |
| Substrate Properties | Peptide-Based Substrate |
| Substrate Type | Kinase Substrate |
| Target Enzyme | Serine/Threonine Kinases |
| Label or Dye | FITC (Fluorescein) |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?