missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein-Crosstide Substrate

Product Code. 10115012
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10115012 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10115012 Supplier Invitrogen Supplier No. PV3509
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Fluorescein-Crosstide Substrate

A general substrate for Ser/Thr kinases, the Crosstide peptide substrate is based upon GSK-3alpha/beta.

For use in combination with the LanthaScreen™ Tb-pCrosstide Antibody (PV3564) to evaluate inhibitors in medium to high throughput screening applications.
TRUSTED_SUSTAINABILITY

Specifications

Amino Acid Sequence Fluorescein- GRPRTSSFAEG
Assay Entry Biochemical terbium phospho-specific activity
Content And Storage 1 mg/mL Fluorescein-Crosstide substrates in 50 mM HEPES, pH 7.5. Store at -20°C and protect from light.
Detection Method Fluorescence
Quantity 1 mg
Substrate Fluorescein-Crosstide
Substrate Properties Peptide-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Serine/Threonine Kinases
Gene ID (Entrez) 10000, 10298, 207, 208, 25400, 51347, 5566, 56164, 57144, 6197, 6198, 8536
Label or Dye FITC (Fluorescein)
Molecular Weight (g/mol) 1524 Da
Research Category Signal Transduction
Shipping Condition Dry Ice
Target Entry PAK4, TAOK3, CAMK2D, CAMK2A, PAK1, PRKACA, AKT2, RPS6KB1, AKT3, AKT1, PAK7, RPS6KA3
Validated Kinases AKT1, AKT2, AKT3, CAMK2A, CAMK2D, PAK1, PAK4, PAK7, PRKACA, RPS6KB1, RPS6KA3, TAOK3
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.