missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fluorescein, disodium salt dihydrate, 90%, pure

Product Code. 10752402 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10752402

Brand: Thermo Scientific Chemicals 410641000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 1264198-62-0 | C20H10Na2O5·2H2O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1264198-62-0
Molecular Formula C20H10Na2O5·2H2O
MDL Number MFCD00150814
Synonym Acid Yellow 73 dihydrate, Fluorescein, water soluble dihydrate, Uranine dihydrate, C.I. 45350 dihydrate

Specifications

Assay Percent Range 90%
Beilstein 19, 222
Merck Index 14, 4159
Solubility Information Solubility in water: soluble.
Infrared Spectrum Authentic
Formula Weight 412.3
Percent Purity 90%
Grade Pure
Packaging Glass bottle
UV_Vis 88% min. (on dry substance)
Color Brown to Orange/Red or Brown/Green
Quantity 100 g
Physical Form Powder
Chemical Name or Material Fluorescein, disodium salt dihydrate, 90%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.