missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein, Free Acid, Reag. Ph. Eur., Solstice

Product Code. 15623820
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic Bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15623820 100 g Plastic Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15623820 Supplier Honeywell-Fluka Supplier No. 32615100G
  • ONLINE EXCLUSIVE PRICE
    74.46 €
    62.67 € / 100g
    Save 11.79 €
    15% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
Visualizzare prodotti alternativi

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Reag. Ph. Eur., free acid

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 2321-07-5
Molecular Formula C20H12O5
Molecular Weight (g/mol) 332.31
MDL Number MFCD00005050
InChI Key GNBHRKFJIUUOQI-UHFFFAOYSA-N
PubChem CID 16850
ChEBI CHEBI:31624
IUPAC Name 3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O

Specifica

Melting Point 314°C to 316°C
UN Number NONH for all modes of transport
Beilstein 94324
Formula Weight 332.31g/mol
Odor Odourless
Packaging Plastic Bottle
Suitability Passes test for UV spectrum
Color Index No. Acid Yellow 73
Color Brown
Quantity 100 g
Chemical Name or Material Fluorescein, Reag. Ph. Eur.
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.