missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fluorescein isothiocyanate, isomer 1, 95%

Product Code. 11327346
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327346 1 g 1g
11337346 50 mg 50mg
11347346 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11327346 Supplier Thermo Scientific Chemicals Supplier No. L09319.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3326-32-7 | C21H11NO5S | 389.381 g/mol

Fluorescein isothiocyanate is used as fluorescent labeling reagent for proteins and for rapid identification of pathogens in fluorescent antibody technique. It is also used as a reagent for microsequencing of proteins and peptides (HPLC). On celite it is used for the labeling of fibrinogen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorescein isothiocyanate is used as fluorescent labeling reagent for proteins and for rapid identification of pathogens in fluorescent antibody technique. It is also used as a reagent for microsequencing of proteins and peptides (HPLC). On celite it is used for the labeling of fibrinogen.

Solubility
Slightly soluble in DMSO, ethanol, methanol DMF and acetone

Notes
Incompatible with strong oxidizing agents, strong bases and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3326-32-7
Molecular Formula C21H11NO5S
Molecular Weight (g/mol) 389.381
MDL Number MFCD00005063
InChI Key MHMNJMPURVTYEJ-UHFFFAOYSA-N
Synonym fitc, fluorescein isothiocyanate isomer i, 5-isothiocyanatofluorescein, fluorescein 5-isothiocyanate, 5-fitc, fluorescein-5-isothiocyanate, fluorescein 5 6-isothiocyanate, fitc isomer 1, unii-i223nx31w9, fitc isomer
PubChem CID 18730
ChEBI CHEBI:37918
IUPAC Name 3',6'-dihydroxy-6-isothiocyanatospiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC2=C(C=C1N=C=S)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O

Specifications

Melting Point >360°C
Quantity 1 g
Beilstein 1407295
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in DMSO,ethanol,methanol DMF and acetone
Formula Weight 389.39
Percent Purity 95%
Chemical Name or Material Fluorescein isothiocyanate, isomer 1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.