missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein, Laser Grade 99%, Thermo Scientific Chemicals

Product Code. 10307771 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10307771 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10307771 Supplier Thermo Scientific Acros Supplier No. 410620010
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2321-07-5 | Molecular Formula: C20H12O5 | Molecular Weight: 332.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2321-07-5
Molecular Formula C20H12O5
Molecular Weight (g/mol) 332.31
InChI Key GNBHRKFJIUUOQI-UHFFFAOYSA-N
PubChem CID 16850
ChEBI CHEBI:31624
IUPAC Name 3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O

Specifications

Melting Point 314.0°C to 316.0°C
Fluorescence Max 518nm (in basic CH3OH)
Infrared Spectrum Authentic
Lambda Max 498nm (in basic MeOH)
Molar Absorption Coefficient 63900L/(mole.cm) (lambda max.)
Packaging Glass bottle
Suitability (for use as a laser dye) passes
Assay Percent Range 99%
Beilstein 19, 222
Merck Index 15, 4192
Color Orange to Red
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dilute alkalies and dilute acids, soluble in boiling alcohol and ether, insoluble in benzene and chloroform
Quantity 1 g
Physical Form Powder
Formula Weight 332.3
Percent Purity 99%
Grade Pure
Chemical Name or Material Fluorescein, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Fluorescein
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Fluorescein

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.