missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein-MAP2K1

Product Code. 10144603
Change view
Click to view available options
Quantity:
20 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10144603 20 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10144603 Supplier Invitrogen Supplier No. PV4812
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Fluorescein-MAP2K1

Inactive MAP2K1 (MEK1) has been labeled with fluorescein, and is a substrate for MAPK family members.

For use in combination with the LanthaSreen™ Tb-MAP2K1 [pSer217/221] Antibody (PV4813) to evaluate inhibitors in medium to high throughput screening applications.
TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Biochemical terbium phospho-specific activity
Content And Storage Fluorescein-MAP2K1 protein substrate in 10 mM Tris, pH 7.5, 150 mM NaCl, 10% glycerol, 1mM DTT. Store at -80°C.
Detection Method Fluorescence
Quantity 20 nmol
Substrate MAP2K1
Substrate Properties Protein-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Serine/Threonine Kinases
Gene ID (Entrez) 10454, 10746, 1326, 4215, 4217, 4293, 4294, 4296, 5894, 673, 6885
Label or Dye FITC (Fluorescein)
Molecular Weight (g/mol) 50000 Da
Shipping Condition Dry Ice
Target Entry MAP3K2, RAF1 (cRAF) Y340D Y341D, MAP3K10, MAP3K9, MAP3K7/MAP3K7IP1, MAP3K5, MAP3K11, BRAF V599E, BRAF, MAP3K8, MAP3K3
Validated Kinases BRAF, MAP3K2, MAP3K3, MAP3K5, MAP3K7, MAP3K8, MAP3K9, MAP3K10, MAP3K11
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.