missing translation for 'onlineSavingsMsg'
Learn More

Fluorescein-P53 (Ser15) Peptide Substrate

Product Code. 10401805
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10401805 1 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10401805 Supplier Invitrogen Supplier No. PV5132

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluorescein-p53 (Ser15) Peptide Substrate

A fluorescein-labeled peptide substrate based upon p53 containing Ser15. p53 is phosphorylated at Ser15 by a wide range of kinases including PI3K/AKT pathway kinases such DNA-PK, ATM, and ATR.

For use in combination with the LanthaScreen™ Tb-anti-phospho-p53 [pSer15] antibody (PV5130) to evaluate inhibitors in medium to high throughput screening applications.
TRUSTED_SUSTAINABILITY

Specifications

Amino Acid Sequence Fluorescein-EPPLSQEAFADLWKK
Assay Entry Biochemical terbium phospho-specific activity
Content And Storage 1 mg/mL Fluorescein-p53 (Ser15) peptide substrates in DMSO. Store at -20° C and protect from light.
Detection Method Fluorescence
Product Line LanthaScreen
Quantity 1 mL
Substrate Fluorescein-Tb
Substrate Properties Peptide-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Serine/Threonine Kinases
Label or Dye FITC (Fluorescein)
Molecular Weight (g/mol) 2118.3 Da
Shipping Condition Dry Ice
Target Entry DNAPK
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.