missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fluorescein-PKC Substrate

Product Code. 10319082
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10319082 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10319082 Supplier Thermo Scientific™ Supplier No. PV3506

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A fluorescein-labeled peptide substrate for the Protein Kinase C (PKC) family.

A fluorescein-labeled peptide substrate for the Protein Kinase C (PKC) family.

For use in combination with the LanthaScreen™ Tb-pSer (PKC Substrate) antibody (Cat. No. PV3560) to evaluate inhibitors in medium to high throughput screening applications.

TRUSTED_SUSTAINABILITY

Specifications

Amino Acid Sequence Fluorescein- RFARKGSLRQKNV
Assay Entry Biochemical terbium phospho-specific activity
Chromagen Quantity 1mg
Content And Storage 1 mg/mL Fluorescein PKC peptide substrates in water. Store at -20°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Pharma and biopharma, protein assays and analysis, target and lead identification and validation, combination with the LanthaScreen™ Tb-pSer (PKC Substrate) antibody (PV3560) to evaluate inhibitors in medium to high throughput screening applications,
Quantity 1 mg
Substrate Fluorescein-PKC
Substrate Properties Peptide-Based Substrate
Substrate Type Kinase Substrate
Target Enzyme Serine/Threonine Kinases
Item Description Fluorescein-PKC Substrate, 1mg, 1mg/mL Concentration, 1919Da Molecular Weight, A Fluorescein-labeled Peptide Substrate For the Protein Kinase C (PKC) Family
Label or Dye FITC (Fluorescein)
Molecular Weight (g/mol) 1919 Da
Product Type Fluorescein-PKC Substrate
Shipping Condition Dry Ice
Storage Requirements –20°C
Substrate Quantity 1mg
Target Entry MAP4K5, PRKCH, PRKCQ, PRKX, ROCK2, PRKCD, NEK2, PRKCG, SGK, MAP4K2, MINK1, SGK2, PRKCA, PRKCI, AURKB, TAOK2, MAP4K4, MARK4, DYRK4, PHKG1, PRKCZ, ROCK1, RPS6KA1, PRKCB1, STK24, PRKCB2, RPS6KA2, SGKL, STK25, PRKCE, MST4
Validated Kinases AURKB, DYRK4, MARK4, MAP4K2, MAP4K4, MAP4K5, MINK1, MST4, NEK2, PHKG1, PKC, PRKX, ROCK2, RPS6KA1, RPS6KA2, STK24, STK25, SGK, SGK2, SGKL, TAOK2, TBK1
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.