missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Fluorescent Brightener 28

Product Code. 11445501 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445501 1 g 1g
11465501 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445501 Supplier MP Biomedicals™ Supplier No. 0215806701
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Fluoresces clear bluish under UV radiation. Fluorescent Brightener 28 is a colorless organic compound that is used for the staining of fungi and as a viability stain. Industrial uses include use as a fluorescent brightening agent for cellulose and polyamide fabrics, paper and detergents and soaps.

  • This product is related structurally to Fluorescent Brightner 9 (Calcofluor White MR), which was used by Jefferies and Belcher and by Maeda and Ischida in plant tissue studies.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 g
Storage Requirements Store at Room Temperature (15°C to 30°C)
Format Powder
Concentration 0.05% to 0.20%
For Use With (Application) Fluorescent Brightener 28 is used for the staining of fungi and as a viability stain. Industrial uses include use as a fluorescent brightening agent for cellulose and polyamide fabrics, paper and in detergents and soaps.
Color Yellow
Melting Point 290°C
CAS 4404-43-7
Molecular Formula C40H42N12O10S2Na2
Formula Weight 960.96
Synonym Calcofluor White M2R, Tinopal UNPA-GX
Product Type Fluorescent Brightener
Show More Show Less

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.